missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzo[b]furan-2-carboxylic acid, 97%, Thermo Scientific Chemicals

Product Code. 11429360
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10242-10-1
Molecular Formula C9H4ClO3
Molecular Weight (g/mol) 195.58
MDL Number MFCD00060512
InChI Key JETRXAHRPACNMA-UHFFFAOYSA-M
Synonym 5-chlorobenzofuran-2-carboxylic acid, 5-chlorobenzo b furan-2-carboxylic acid, 5-chloro-benzofuran-2-carboxylic acid, 2-benzofurancarboxylic acid, 5-chloro, 2-benzofurancarboxylicacid, 5-chloro, 2-carboxy-5-chlorobenzo b furan, 5-chloro-2-benzofurancarboxylic acid, 5-chlorocoumarilic acid, acmc-20am44, 5-chlo-robenzofuran-2-carboxylic acid
PubChem CID 937838
IUPAC Name 5-chloro-1-benzofuran-2-carboxylic acid
SMILES [O-]C(=O)C1=CC2=CC(Cl)=CC=C2O1

Specifications

Quantity 5 g
Formula Weight 196.59
Percent Purity 97%
Chemical Name or Material 5-Chlorobenzo[b]furan-2-carboxylic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.