Learn More
5-Chlorobenzo[b]thiophene, 97%, Thermo Scientific Chemicals
208.00 € - 811.00 €
Chemical Identifiers
CAS | 20532-33-6 |
---|---|
Molecular Formula | C8H5ClS |
Molecular Weight (g/mol) | 168.638 |
MDL Number | MFCD00055623 |
InChI Key | SNYURIHMNFPQFL-UHFFFAOYSA-N |
Synonym | 5-chlorobenzothiophene, 5-chlorobenzo b thiophene, 5-chloro-benzo b thiophene, 5-chloro benzothiophene, benzo b thiophene, 5-chloro, zlchem 66, 5-chlorothionaphthene, 5-chloranyl-1-benzothiophene, benzo b thiophene,5-chloro, ksc544i5d |
PubChem CID | 11309754 |
IUPAC Name | 5-chloro-1-benzothiophene |
SMILES | C1=CC2=C(C=CS2)C=C1Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15488308
|
Thermo Scientific Alfa Aesar
H61748.06 |
5 g |
208.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15498308
|
Thermo Scientific Alfa Aesar
H61748.14 |
25 g | N/A | ||||||
Beschreibung
5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Chlorobenzo[b]thiophene is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Chemische Identifikatoren
20532-33-6 | |
168.638 | |
SNYURIHMNFPQFL-UHFFFAOYSA-N | |
11309754 | |
C1=CC2=C(C=CS2)C=C1Cl |
C8H5ClS | |
MFCD00055623 | |
5-chlorobenzothiophene, 5-chlorobenzo b thiophene, 5-chloro-benzo b thiophene, 5-chloro benzothiophene, benzo b thiophene, 5-chloro, zlchem 66, 5-chlorothionaphthene, 5-chloranyl-1-benzothiophene, benzo b thiophene,5-chloro, ksc544i5d | |
5-chloro-1-benzothiophene |
Spezifikation
20532-33-6 | |
C8H5ClS | |
5-chlorobenzothiophene, 5-chlorobenzo b thiophene, 5-chloro-benzo b thiophene, 5-chloro benzothiophene, benzo b thiophene, 5-chloro, zlchem 66, 5-chlorothionaphthene, 5-chloranyl-1-benzothiophene, benzo b thiophene,5-chloro, ksc544i5d | |
SNYURIHMNFPQFL-UHFFFAOYSA-N | |
5-chloro-1-benzothiophene | |
11309754 | |
97% | |
5-Chlorobenzo[b]thiophene |
White | |
MFCD00055623 | |
Slightly soluble in water. | |
C1=CC2=C(C=CS2)C=C1Cl | |
168.638 | |
168.64 | |
Solid |
Sicherheit und Handhabung
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only