missing translation for 'onlineSavingsMsg'
Learn More

5-Chlorobenzoxazole-2-thiol, 97%, Thermo Scientific™

Product Code. 15495507
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22876-19-3
Molecular Formula C7H4ClNOS
Molecular Weight (g/mol) 185.625
MDL Number MFCD00175241
InChI Key BOBIZYYFYLLRAH-UHFFFAOYSA-N
Synonym 5-chlorobenzo d oxazole-2-thiol, 5-chloro-2-mercaptobenzoxazole, 5-chloro-1,3-benzoxazole-2-thiol, 5-chlorobenzo d oxazole-2 3h-thione, 2-mercapto-5-chlorobenzoxazole, 5-chlorobenzooxazole-2-thiol, 5-chloro-benzooxazole-2-thiol, 2 3h-benzoxazolethione, 5-chloro, 5-chloro-2-thioxobenzoxazoline, 2-benzoxazolethiol, 5-chloro
PubChem CID 708870
IUPAC Name 5-chloro-3H-1,3-benzoxazole-2-thione
SMILES C1=CC2=C(C=C1Cl)NC(=S)O2

Specifications

Melting Point 280°C to 285°C
Quantity 5 g
Formula Weight 185.63
Percent Purity 97%
Chemical Name or Material 5-Chlorobenzoxazole-2-thiol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'