Learn More
5-Chloropyridine-3-boronic acid, 95%, Thermo Scientific™
137.00 € - 1233.00 €
Chemical Identifiers
CAS | 872041-85-5 |
---|---|
Molecular Formula | C5H5BClNO2 |
Molecular Weight (g/mol) | 157.36 |
MDL Number | MFCD06798243 |
InChI Key | NJXYBTMCTZAUEE-UHFFFAOYSA-N |
Synonym | 5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397 |
PubChem CID | 20111877 |
IUPAC Name | (5-chloropyridin-3-yl)boronic acid |
SMILES | OB(O)C1=CC(Cl)=CN=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15403777
|
Thermo Scientific Alfa Aesar
H27756.MD |
250 mg |
137.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11322919
|
Thermo Scientific Alfa Aesar
H27756.03 |
1 g | N/A | ||||||
11332919
|
Thermo Scientific Alfa Aesar
H27756.06 |
5 g | N/A | ||||||
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
872041-85-5 | |
157.36 | |
NJXYBTMCTZAUEE-UHFFFAOYSA-N | |
20111877 | |
OB(O)C1=CC(Cl)=CN=C1 |
C5H5BClNO2 | |
MFCD06798243 | |
5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397 | |
(5-chloropyridin-3-yl)boronic acid |
Specifications
872041-85-5 | |
MFCD06798243 | |
NJXYBTMCTZAUEE-UHFFFAOYSA-N | |
(5-chloropyridin-3-yl)boronic acid | |
20111877 | |
95% |
C5H5BClNO2 | |
5-chloropyridine-3-boronic acid, 5-chloropyridin-3-yl boronic acid, 5-chloro-3-pyridineboronic acid, 3-chloropyridine-5-boronic acid, 5-chloro-3-pyridinyl boronic acid, 5-chloro-3-pyridyl boronic acid, 5-chloropyridin-3-yl-3-boronic acid, 3-chloro-5-pyridyl boronic acid, 5-chloro-3-pyridineboronicacid, pubchem17397 | |
OB(O)C1=CC(Cl)=CN=C1 | |
157.36 | |
157.37 | |
5-Chloropyridine-3-boronic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.