Learn More
5-Chlorovaleryl chloride, 96%, Thermo Scientific Chemicals
93.06 € - 754.00 €
Chemical Identifiers
CAS | 1575-61-7 |
---|---|
Molecular Formula | C5H8Cl2O |
Molecular Weight (g/mol) | 155.02 |
MDL Number | MFCD00000758 |
InChI Key | SVNNWKWHLOJLOK-UHFFFAOYSA-N |
Synonym | 5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1 |
PubChem CID | 74089 |
IUPAC Name | 5-chloropentanoyl chloride |
SMILES | ClCCCCC(Cl)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10429072
|
Thermo Scientific Acros
154770100 |
10 mL | Glass bottle |
93.06 €
10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10061760
|
Thermo Scientific Acros
154770250 |
25 mL | Glass bottle |
195.98 €
25mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10123110
|
Thermo Scientific Acros
154771000 |
100 mL | Glass bottle |
754.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1575-61-7 | |
155.02 | |
SVNNWKWHLOJLOK-UHFFFAOYSA-N | |
74089 | |
ClCCCCC(Cl)=O |
C5H8Cl2O | |
MFCD00000758 | |
5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1 | |
5-chloropentanoyl chloride |
Specifications
1575-61-7 | |
100.0 | |
1.2000g/mL | |
Authentic | |
C5H8Cl2O | |
1.4630 to 1.4650 | |
1.2 | |
Solubility in water: hydrolyses | |
ClCCCCC(Cl)=O | |
155.02 | |
155.02 | |
Liquid |
95.0 | |
Colorless to Yellow | |
90°C | |
95% min. (GC) | |
Cl(CH2)4COCl | |
MFCD00000758 | |
5-chlorovaleryl chloride, valeryl chloride, 5-chloro, pentanoyl chloride, 5-chloro, 5-chlorovalerylchloride, 5-chlorovaleroyl chloride, pentanoylchloride, 5-chloro, chlorovaleryl chloride, pubchem10919, 5-chloro-pentanoylchloride, acmc-1cuj1 | |
SVNNWKWHLOJLOK-UHFFFAOYSA-N | |
5-chloropentanoyl chloride | |
74089 | |
96% | |
5-Chlorovaleryl chloride, 96% |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 216-403-1
missing translation for 'rtecsNumber' : YV9108000
- 5-Chlorovaleryl chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only