missing translation for 'onlineSavingsMsg'
Learn More

5alpha-Cholestane, 98+%

Product Code. 10327300 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10327300

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 481-21-0 | C27H48 | 372.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-21-0
Molecular Formula C27H48
Molecular Weight (g/mol) 372.67
MDL Number MFCD00066412
InChI Key XIIAYQZJNBULGD-XWLABEFZSA-N
Synonym 5alpha-cholestane, 5-alpha-cholestane, alpha-cholestane, cholestane, 28,29,30-trinorlanostane, unii-u260hwn305, 5alpha-cholestane 8ci, 5?-cholestane, cholestane van
PubChem CID 2723895
ChEBI CHEBI:35515
IUPAC Name (5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCCC4)C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 78°C to 81°C
Color White
Boiling Point 250°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 250 mg
Merck Index 15, 2206
Specific Rotation + 26.5
Solubility Information Solubility in water: insoluble in water. Other solubilities: freely soluble in chloroform (50mg/mL),ether and,slightly soluble in absolute alcohol
Formula Weight 372.67
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 5α-Cholestane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.