missing translation for 'onlineSavingsMsg'
Learn More

5α-Cholestane, 98+%, Thermo Scientific Chemicals

Product Code. 10459080
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 481-21-0
Molecular Formula C27H48
Molecular Weight (g/mol) 372.67
MDL Number MFCD00066412
InChI Key XIIAYQZJNBULGD-XWLABEFZSA-N
Synonym 5alpha-cholestane, 5-alpha-cholestane, alpha-cholestane, cholestane, 28,29,30-trinorlanostane, unii-u260hwn305, 5alpha-cholestane 8ci, 5?-cholestane, cholestane van
PubChem CID 2723895
ChEBI CHEBI:35515
IUPAC Name (5R,8R,9S,10S,13R,14S,17R)-10,13-dimethyl-17-[(2R)-6-methylheptan-2-yl]-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthrene
SMILES CC(C)CCCC(C)C1CCC2C1(CCC3C2CCC4C3(CCCC4)C)C

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point 78°C to 81°C
Color White
Boiling Point 250°C (1.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Quantity 1 g
Merck Index 15, 2206
Specific Rotation + 26.5
Solubility Information Solubility in water: insoluble in water. Other solubilities: freely soluble in chloroform (50mg/mL),ether and,slightly soluble in absolute alcohol
Formula Weight 372.67
Percent Purity 98+%
Physical Form Crystalline Powder
Chemical Name or Material 5α-Cholestane
Show More Show Less
Product Identifier
  • 5-a-Cholestane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'