missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Infrared Spectrum | Authentic |
Assay Percent Range | 96% min. (HPLC) |
Quantity | 5 g |
Formula Weight | 125.13 |
Percent Purity | 97% |
Chemical Name or Material | 5-Cyclopropyl-1,2,4-oxadiazol-3-amine |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-5 of 8
Safety and Handling

Product Identifier
- 5-Cyclopropyl-1,2,4-oxadiazol-3-amine
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
5-Cyclopropyl-1,2,4-oxadiazol-3-amine, 97%, Thermo Scientific™ >
Spot an opportunity for improvement?Share a Content Correction