missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5'-Deoxy-5-fluorocytidine, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
Specifications
Molecular Weight (g/mol) | 245.21 |
Purity | ≥94 % |
Quantity | 1 g |
Solubility Information | Miscible with methanol, DMSO |
Color | White to almost white |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction