Learn More
Applied Biosystems™ 5' Fluorescent Labeled Oligo
5' Fluorescent Labeled Oligo
Brand: Applied Biosystems™ 450017H
This item is not returnable.
View return policy
Description
Custom 5'-labeled primers are ideal for applications including fragment analysis, gene expression studies, genotyping, and PCR. Choose among FAM™, TET™, VIC™, HEX™, NED™, and PET™ as fluorescent 5' labels. Oligos are desalted; HPLC purification is also available.Specifications
5' Fluorescent Labeled Single Primer, 300 kpmol | |
1.524 m | |
Desalted, HPLC | |
5' Labeled | |
HPLC |
5' Fluorescent Labeled Single Primer | |
Store in Freezer (-5°C to -30°C). | |
FAM, HEX, NED, PET, TET, VIC | |
300,000 pmol | |
Fragment Analysis, Gene Expression Studies, Genotyping, PCR |