missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoro-2-methoxybenzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15475507
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179897-94-0
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.95
MDL Number MFCD01863526
InChI Key CCQKIRUMTHHPSX-UHFFFAOYSA-N
Synonym 5-fluoro-2-methoxyphenyl boronic acid, 2-methoxy-5-fluorophenylboronic acid, 5-fluoro-2-methoxybenzeneboronic acid, 2-methoxy-5-fluorobenzeneboronic acid, 5-fluoro-2-methoxyphenylboronicacid, 2-borono-4-fluoroanisole, 3-fluoro-6-methoxybenzeneboronic acid, 5-fluoro-2-methoxyphenyl boranediol, 4-fluoroanisole-2-boronic acid, boronic acid, 5-fluoro-2-methoxyphenyl
PubChem CID 2782673
IUPAC Name (5-fluoro-2-methoxyphenyl)boronic acid
SMILES COC1=CC=C(F)C=C1B(O)O

Specifications

Quantity 25 g
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 5-Fluoro-2-methoxybenzeneboronic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.