missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Fluoroindole-2-carboxylic acid, 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
5-Fluoroindole-2-carboxylic acid,is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.
Solubility
It is highly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents. Stable under normal temperatures and pressures.
Specifications
Specifications
Melting Point | ∼259°C (decomposition) |
Quantity | 5 g |
Beilstein | 153217 |
Solubility Information | It is highly soluble in water. |
Formula Weight | 179.15 |
Percent Purity | ≥98% |
Chemical Name or Material | 5-Fluoroindole-2-carboxylic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'