missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole-3-carboxaldehyde, 98%, Thermo Scientific Chemicals

47.40 € - 516.00 €

Chemical Identifiers

CAS 2338-71-8
Molecular Formula C9H6FNO
Molecular Weight (g/mol) 163.15
MDL Number MFCD00022719
InChI Key YUAJKGBLPVLADK-UHFFFAOYSA-N
Synonym 5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
PubChem CID 259089
IUPAC Name 5-fluoro-1H-indole-3-carbaldehyde
SMILES FC1=CC=C2NC=C(C=O)C2=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11348466
View Documents
Thermo Scientific Alfa Aesar
L17507.MD
250 mg
47.40 €
250mg
Please to purchase this item. Need a web account? Register with us today!
11328466
View Documents
Thermo Scientific Alfa Aesar
L17507.03
1 g
133.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
11338466
View Documents
Thermo Scientific Alfa Aesar
L17507.06
5 g
516.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2338-71-8
163.15
YUAJKGBLPVLADK-UHFFFAOYSA-N
259089
FC1=CC=C2NC=C(C=O)C2=C1
C9H6FNO
MFCD00022719
5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
5-fluoro-1H-indole-3-carbaldehyde

Specifications

2338-71-8
C9H6FNO
1449216
5-fluoroindole-3-carboxaldehyde, 5-fluoro-1h-indole-3-carboxaldehyde, 5-fluoroindole-3-carbaldehyde, 1h-indole-3-carboxaldehyde, 5-fluoro, 3-formyl-5-fluoroindole, pubchem7267, zlchem 1197, 5-fluoro-3-formylindole, 5-fluoroindole-3-aldehyde, acmc-209g2v
FC1=CC=C2NC=C(C=O)C2=C1
163.15
163.15
5-Fluoroindole-3-carboxaldehyde
164°C
MFCD00022719
Air Sensitive
YUAJKGBLPVLADK-UHFFFAOYSA-N
5-fluoro-1H-indole-3-carbaldehyde
259089
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only