missing translation for 'onlineSavingsMsg'
Learn More

5-Fluoroindole, 98%, Thermo Scientific Chemicals

29.35 € - 677.37 €

Chemical Identifiers

CAS 399-52-0
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00005671
InChI Key ODFFPRGJZRXNHZ-UHFFFAOYSA-N
Synonym 5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
PubChem CID 67861
ChEBI CHEBI:72818
IUPAC Name 5-fluoro-1H-indole
SMILES FC1=CC=C2NC=CC2=C1
View More Specs

Products 4
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10397521
View Documents
Thermo Scientific
311922500
250 mg Glass bottle
29.35 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10183961
View Documents
Thermo Scientific Acros
311920010
1 g Glass bottle
56.08 €
1g
Please to purchase this item. Need a web account? Register with us today!
13448508
View Documents
Thermo Scientific Acros
311920050
5 g Glass bottle
167.84 €
5g
Please to purchase this item. Need a web account? Register with us today!
13458508
View Documents
Thermo Scientific Acros
311920250
25 g Glass bottle
677.37 €
25g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

399-52-0
135.14
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
67861
5-fluoro-1H-indole
C8H6FN
MFCD00005671
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
CHEBI:72818
FC1=CC=C2NC=CC2=C1

Specifications

399-52-0
100.0
White
Authentic
C8H6FN
5-fluoroindole, 1h-indole, 5-fluoro, 5-fluoro indole, indole, 5-fluoro, 5-fluoro-indole, 5-fluor-1h-indol, 5-fluoroindol, 5-fluoro-indol, 3fuf, pubchem1681
FC1=CC=C2NC=CC2=C1
135.14
CHEBI:72818
98%
5-Fluoroindole, 98%
97.5
44.0°C to 48.0°C
>110°C
97.5% min. (GC)
MFCD00005671
ODFFPRGJZRXNHZ-UHFFFAOYSA-N
5-fluoro-1H-indole
67861
135.14
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 206-917-4

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.