Learn More
5-Fluoroindoline, 97%, Thermo Scientific Chemicals
39.80 € - 389.94 €
Chemical Identifiers
CAS | 2343-22-8 |
---|---|
Molecular Formula | C8H8FN |
Molecular Weight (g/mol) | 137.157 |
MDL Number | MFCD00214461 |
InChI Key | NXQRMQIYCWFDGP-UHFFFAOYSA-N |
PubChem CID | 2774463 |
IUPAC Name | 5-fluoro-2,3-dihydro-1H-indole |
SMILES | C1CNC2=C1C=C(C=C2)F |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15440528
|
Thermo Scientific Alfa Aesar
H54158.MD |
250 mg |
39.80 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15420528
|
Thermo Scientific Alfa Aesar
H54158.03 |
1 g |
100.50 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15430528
|
Thermo Scientific Alfa Aesar
H54158.06 |
5 g |
389.94 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
5-Fluoroindoline is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications5-Fluoroindoline is used as pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Air sensitive. Incompatible with oxidizing agents and heat.
Chemical Identifiers
2343-22-8 | |
137.157 | |
NXQRMQIYCWFDGP-UHFFFAOYSA-N | |
5-fluoro-2,3-dihydro-1H-indole |
C8H8FN | |
MFCD00214461 | |
2774463 | |
C1CNC2=C1C=C(C=C2)F |
Specifications
2343-22-8 | |
MFCD00214461 | |
5-fluoroindoline, 5-fluoro-2,3-dihydro-1h-indole, 1h-indole,5-fluoro-2,3-dihydro, 1h-indole, 5-fluoro-2,3-dihydro, 5-fluoro-indoline, peakdale1_000398, 5-fluoro-2,3-dihydro-1h-indol, 2,3-dihydro-5-fluoro-1h-indole | |
NXQRMQIYCWFDGP-UHFFFAOYSA-N | |
5-fluoro-2,3-dihydro-1H-indole | |
2774463 | |
97% |
C8H8FN | |
Air sensitive | |
Slightly soluble in water. | |
C1CNC2=C1C=C(C=C2)F | |
137.157 | |
137.16 | |
5-Fluoroindoline |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Keep cold; Store under Argon
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.