missing translation for 'onlineSavingsMsg'
Learn More

5-Hydroxyanthranilic acid, 99%, Thermo Scientific Chemicals

Product Code. 10149793
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 394-31-0
Molecular Formula C7H7NO3
Molecular Weight (g/mol) 153.14
MDL Number MFCD00007870
InChI Key HYNQTSZBTIOFKH-UHFFFAOYSA-N
Synonym 5-hydroxyanthranilic acid, diabeton, benzoic acid, 2-amino-5-hydroxy, 2-amino-5-hydroxy benzoic acid, unii-uf9ww3i410, 2-amino-5-hydroxy-benzoic acid, 5-hydroxyanthranillicacid, pubchem15307, 4-amino-3-carboxyphenol, 5-hydroxyanthranillic acid
PubChem CID 164592
IUPAC Name 2-amino-5-hydroxybenzoic acid
SMILES C1=CC(=C(C=C1O)C(=O)O)N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 233°C to 234°C
Color Brown-Gray
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula HOC6H3(NH2)CO2H
Quantity 1 g
Solubility Information Solubility in water: insoluble
Formula Weight 153.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 5-Hydroxyanthranilic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'