missing translation for 'onlineSavingsMsg'
Learn More
5-Hydroxyisophthalic acid, 98%, Thermo Scientific Chemicals Product Code.: 11471498

5-Hydroxyisophthalic acid, 98%, Thermo Scientific Chemicals

Product Code. 11471498
50 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11471498

Brand: Thermo Scientific Alfa Aesar A13845.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-83-7
Molecular Formula C8H6O5
Molecular Weight (g/mol) 182.131
MDL Number MFCD00002515
InChI Key QNVNLUSHGRBCLO-UHFFFAOYSA-N
Synonym 5-hydroxyisophthalic acid, 5-oxyisophthalic acid, isophthalic acid, 5-hydroxy, 1,3-benzenedicarboxylic acid, 5-hydroxy, 5-hydoxyisophthalic acid, 5-hydroxy-isophthalic acid, 5-hydroxy-1,3-benzenedicarboxylic acid, 5-hydroxyisosphthalic acid, isophthalic acid, 5-hydroxy-6ci,7ci,8ci, 5-hydroxyisophthalicacidShow MoreShow Less
PubChem CID 69257
IUPAC Name 5-hydroxybenzene-1,3-dicarboxylic acid
SMILES C1=C(C=C(C=C1C(=O)O)O)C(=O)O

Specifications

Melting Point ∼300°C (decomposition)
Odor Odorless
Quantity 50 g
Beilstein 2693561
Formula Weight 182.13
Percent Purity 98%
Chemical Name or Material 5-Hydroxyisophthalic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10246797 Certificate TypeCertificate of Analysis Date06/06/2023 Catalog NumberA13845.18
Lot Number10240059 Certificate TypeCertificate of Analysis Date15/04/2022 Catalog NumberA13845.18,A13845.0B,A13845.30
Lot Number10239825 Certificate TypeCertificate of Analysis Date12/04/2022 Catalog NumberA13845.18,A13845.0B,A13845.30
Lot Number10125114 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13845.18,A13845.0B,A13845.30
Lot Number10147952 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13845.18,A13845.0B,A13845.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Hydroxyisophthalic acid, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.