Learn More
5-Indolylboronic acid, 97%, Thermo Scientific Chemicals
25.13 € - 102.51 €
Chemical Identifiers
CAS | 144104-59-6 |
---|---|
Molecular Formula | C8H8BNO2 |
Molecular Weight (g/mol) | 160.97 |
MDL Number | MFCD01319013 |
InChI Key | VHADYSUJZAPXOW-UHFFFAOYSA-N |
Synonym | 5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid |
PubChem CID | 2734361 |
IUPAC Name | 1H-indol-5-ylboronic acid |
SMILES | OB(O)C1=CC=C2NC=CC2=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10078841
|
Thermo Scientific Acros
359461000 |
100 mg | Glass Bottle |
25.13 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10621843
|
Thermo Scientific Acros
359465000 |
500 mg | Glass bottle |
60.40 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10563884
|
Thermo Scientific Acros
359460010 |
1 g | Glass bottle |
102.51 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
144104-59-6 | |
160.97 | |
VHADYSUJZAPXOW-UHFFFAOYSA-N | |
2734361 | |
OB(O)C1=CC=C2NC=CC2=C1 |
C8H8BNO2 | |
MFCD01319013 | |
5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid | |
1H-indol-5-ylboronic acid |
Specifications
144104-59-6 | |
100.0 | |
Authentic | |
C8H8BNO2 | |
5-indolylboronic acid, indole-5-boronic acid, 1h-indol-5-yl boronic acid, 1h-indole-5-boronic acid, 5-indolboronic acid, 5-indolyl boronic acid, 5-indole-boronic acid, 1h-indol-5-ylboranediol, 5-indole boric acid, 5-indoleboronic acid | |
OB(O)C1=CC=C2NC=CC2=C1 | |
160.97 | |
160.96 | |
Crystalline Powder |
96.0 | |
White | |
96% min. (HPLC) | |
MFCD01319013 | |
VHADYSUJZAPXOW-UHFFFAOYSA-N | |
1H-indol-5-ylboronic acid | |
2734361 | |
97% | |
5-Indolylboronic acid |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician if you feel unwell.
Wear pr
GHS Signal Word: Warning
- 5-Indolylboronic acid
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H312-Harmful in contact with skin.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.