missing translation for 'onlineSavingsMsg'
Learn More

5-Methoxymethyl-2-furoic acid, 99%, Thermo Scientific™

80.00 € - 283.00 €

Chemical Identifiers

CAS 1917-60-8
Molecular Formula C7H8O4
Molecular Weight (g/mol) 156.137
MDL Number MFCD04367609
InChI Key HUUCEXDTXVRYMK-UHFFFAOYSA-N
Synonym 5-methoxymethyl furan-2-carboxylic acid, 5-methoxymethyl-furan-2-carboxylic acid, 5-methoxymethyl-2-furoic acid, acmc-1bvdc, sumiki's acid methyl ether, 2-furancarboxylic acid,5-methoxymethyl
PubChem CID 1125227
IUPAC Name 5-(methoxymethyl)furan-2-carboxylic acid
SMILES COCC1=CC=C(O1)C(=O)O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11479470
View Documents
Thermo Scientific Alfa Aesar
H50314.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11489470
View Documents
Thermo Scientific Alfa Aesar
H50314.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

1917-60-8
156.137
HUUCEXDTXVRYMK-UHFFFAOYSA-N
1125227
COCC1=CC=C(O1)C(=O)O
C7H8O4
MFCD04367609
5-methoxymethyl furan-2-carboxylic acid, 5-methoxymethyl-furan-2-carboxylic acid, 5-methoxymethyl-2-furoic acid, acmc-1bvdc, sumiki's acid methyl ether, 2-furancarboxylic acid,5-methoxymethyl
5-(methoxymethyl)furan-2-carboxylic acid

Specifications

1917-60-8
MFCD04367609
HUUCEXDTXVRYMK-UHFFFAOYSA-N
5-(methoxymethyl)furan-2-carboxylic acid
1125227
99%
C7H8O4
5-methoxymethyl furan-2-carboxylic acid, 5-methoxymethyl-furan-2-carboxylic acid, 5-methoxymethyl-2-furoic acid, acmc-1bvdc, sumiki's acid methyl ether, 2-furancarboxylic acid,5-methoxymethyl
COCC1=CC=C(O1)C(=O)O
156.137
156.14
5-Methoxymethyl-2-furoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.