missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-benzothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10443763
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 27035-41-2
Molecular Formula C10H8OS
Molecular Weight (g/mol) 176.233
MDL Number MFCD05865141
InChI Key SIHFZJLPKQUYAH-UHFFFAOYSA-N
Synonym 5-methylbenzo b thiophene-2-carbaldehyde, 5-methyl-benzo b thiophene-2-carbaldehyde, benzo b thiophene-2-carboxaldehyde,5-methyl, 5-methylbenzothiophene-2-carboxaldehyde, 5-methylbenzo b thiophene-2-carboxaldehyde, 5-methyl-1-benzothiophene-2-carboxaldehyde, 5-methyl-1-benzo b thiophene-2-carbaldehyde
PubChem CID 2794770
IUPAC Name 5-methyl-1-benzothiophene-2-carbaldehyde
SMILES CC1=CC2=C(C=C1)SC(=C2)C=O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 176.24
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-benzothiophene-2-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'