missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-1-phenyl-1H-pyrazol-4-yl)methanol, 97%, Thermo Scientific™

29.00 € - 32.20 €

Chemical Identifiers

CAS 153863-35-5
Molecular Formula C11H12N2O
Molecular Weight (g/mol) 188.23
MDL Number MFCD02682046
InChI Key JTNWKZVHKLAHTN-UHFFFAOYSA-N
Synonym 5-methyl-1-phenyl-1h-pyrazol-4-yl methanol, 5-methyl-1-phenylpyrazol-4-yl methanol, 1h-pyrazole-4-methanol,5-methyl-1-phenyl, acmc-1cgp6, 1-phenyl-5-methyl-pyrazole-4-methanol, 5-methyl-1-phenylpyrazol-4-yl methan-1-ol
PubChem CID 2776440
IUPAC Name (5-methyl-1-phenylpyrazol-4-yl)methanol
SMILES CC1=C(CO)C=NN1C1=CC=CC=C1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10661283
View Documents
Thermo Scientific Maybridge
CC15509CB
250 mg Amber Glass Bottle
29.00 €
250mg
Please to purchase this item. Need a web account? Register with us today!
10029141
View Documents
Thermo Scientific Maybridge
CC15509DA
1 g Amber Glass Bottle
32.20 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

153863-35-5
188.23
JTNWKZVHKLAHTN-UHFFFAOYSA-N
2776440
CC1=C(CO)C=NN1C1=CC=CC=C1
C11H12N2O
MFCD02682046
5-methyl-1-phenyl-1h-pyrazol-4-yl methanol, 5-methyl-1-phenylpyrazol-4-yl methanol, 1h-pyrazole-4-methanol,5-methyl-1-phenyl, acmc-1cgp6, 1-phenyl-5-methyl-pyrazole-4-methanol, 5-methyl-1-phenylpyrazol-4-yl methan-1-ol
(5-methyl-1-phenylpyrazol-4-yl)methanol

Specifications

153863-35-5
100.0
C11H12N2O
5-methyl-1-phenyl-1h-pyrazol-4-yl methanol, 5-methyl-1-phenylpyrazol-4-yl methanol, 1h-pyrazole-4-methanol,5-methyl-1-phenyl, acmc-1cgp6, 1-phenyl-5-methyl-pyrazole-4-methanol, 5-methyl-1-phenylpyrazol-4-yl methan-1-ol
CC1=C(CO)C=NN1C1=CC=CC=C1
188.23
188.23
(5-Methyl-1-phenyl-1H-pyrazol-4-yl)methanol
96.5
97%
MFCD02682046
JTNWKZVHKLAHTN-UHFFFAOYSA-N
(5-methyl-1-phenylpyrazol-4-yl)methanol
2776440
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.