missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde, 97%, Thermo Scientific™

Product Code. 11360577
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 98700-50-6
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.214
InChI Key IIEFFVFWBUGUTI-UHFFFAOYSA-N
PubChem CID 2776437
IUPAC Name 5-methyl-1-phenylpyrazole-4-carbaldehyde
SMILES CC1=C(C=NN1C2=CC=CC=C2)C=O

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Synonym 5-methyl-1-phenyl-1h-pyrazole-4-carbaldehyde, 5-methyl-1-phenyl-1h-pyrazole-4-carboxaldehyde, acmc-20am2d, 1h-pyrazole-4-carboxaldehyde, 5-methyl-1-phenyl, 1h-pyrazole-4-carboxaldehyde,5-methyl-1-phenyl
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 5-Methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'