missing translation for 'onlineSavingsMsg'
Learn More

5-Methyl-1H-benzotriazole, 98+%, Thermo Scientific Chemicals

Product Code. 11453314
Click to view available options
Quantity:
25 g
100 g
500 g

5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Methyl-1H-benzotriazole was used in determination of benzothiazoles and benzotriazoles in waste water samples by GC-MS. It is also used as a potential nitrification inhibitor of urea fertilizer in agricultural soils.

Solubility
Soluble in chloroform, methanol, water (6.0 g/L at 25°C), ethanol, and toluene (slightly).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 136-85-6
Molecular Formula C7H7N3
Molecular Weight (g/mol) 133.15
MDL Number MFCD00005702
InChI Key LRUDIIUSNGCQKF-UHFFFAOYSA-N
Synonym 5-methyl-1h-benzotriazole, 5-methylbenzotriazole, tolutriazole, 5-tolyltriazole, 1h-benzotriazole, 5-methyl, 6-methylbenzotriazole, 5-methyl-1h-benzo d 1,2,3 triazole, 5-methyl-1,2,3-benzotriazole, 6-methyl-1,2,3-benzotriazole, t56 bmnnj g1
PubChem CID 8705
ChEBI CHEBI:83455
IUPAC Name 5-methyl-2H-benzotriazole
SMILES CC1=CC2=NNN=C2C=C1

Specifications

Melting Point 77°C to 84°C
pH 5.3
Boiling Point 210°C to 212°C (12 mmHg)
Odor Odorless
Quantity 100 g
Beilstein 116658
Solubility Information Soluble in chloroform,methanol,water (6.0g/L at 25°C),ethanol,and toluene (slightly).
Formula Weight 133.15
Percent Purity ≥98%
Chemical Name or Material 5-Methyl-1H-benzotriazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'