missing translation for 'onlineSavingsMsg'
Learn More

(5-Methyl-2-phenyl-3-furyl)methanol, 95%, Thermo Scientific™

Product Code. 10708075
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 183210-33-5
Molecular Formula C12H12O2
Molecular Weight (g/mol) 188.226
MDL Number MFCD08435905
InChI Key VZFFMWAZPODJRX-UHFFFAOYSA-N
Synonym 5-methyl-2-phenyl-3-furyl methanol, 5-methyl-2-phenylfuran-3-yl methanol, 3-furanmethanol,5-methyl-2-phenyl, 5-methyl-2-phenyl-3-furyl methan-1-ol
PubChem CID 15269738
IUPAC Name (5-methyl-2-phenylfuran-3-yl)methanol
SMILES CC1=CC(=C(O1)C2=CC=CC=C2)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 188.22
Percent Purity 95%
Chemical Name or Material (5-Methyl-2-phenyl-3-furyl)methanol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'