missing translation for 'onlineSavingsMsg'
Learn More
5-Nitro-1H-indazole, 98+%, Thermo Scientific Chemicals Product Code.: 11475553

5-Nitro-1H-indazole, 98+%, Thermo Scientific Chemicals

Product Code. 11475553
100 g
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy

Product Code. 11475553

Brand: Thermo Scientific Alfa Aesar A13478.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5401-94-5
Molecular Formula C7H5N3O2
Molecular Weight (g/mol) 163.136
MDL Number MFCD00005693
InChI Key WSGURAYTCUVDQL-UHFFFAOYSA-N
Synonym 5-nitroindazole, 1h-indazole, 5-nitro, unii-235y7p37zd, ccris 4134, 2h-indazole, 5-nitro, 5ni, 5-nitroindazol, 5-nitro indazole, 5-nitro-indazole, 5-nitro-1h-indazol
PubChem CID 21501
IUPAC Name 5-nitro-1H-indazole
SMILES C1=CC2=C(C=C1[N+](=O)[O-])C=NN2

Specifications

Melting Point 208°C to 211°C
Odor Odorless
Quantity 100 g
Beilstein 7936
Formula Weight 163.14
Percent Purity ≥98%
Chemical Name or Material 5-Nitro-1H-indazole

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10253254 Certificate TypeCertificate of Analysis Date09/12/2024 Catalog NumberA13478.22,A13478.14,A13478.36
Lot Number10249112 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog NumberA13478.22,A13478.14,A13478.36
Lot Number10243504 Certificate TypeCertificate of Analysis Date23/09/2022 Catalog NumberA13478.22,A13478.14,A13478.36
Lot Number10235754 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13478.22,A13478.14,A13478.36
Lot NumberI16M23 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13478.22,A13478.14,A13478.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Nitro-1H-indazole, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.