missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Nitro-2-furaldehyde diacetate, 98%, Thermo Scientific Chemicals
32.50 € - 466.00 €
Chemical Identifiers
CAS | 92-55-7 |
---|---|
Molecular Formula | C9H9NO7 |
Molecular Weight (g/mol) | 243.17 |
MDL Number | MFCD00003244 |
InChI Key | HSXKWKJCZNRMJO-UHFFFAOYSA-N |
Synonym | 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate |
PubChem CID | 7097 |
IUPAC Name | [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate |
SMILES | CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10791431
|
Thermo Scientific Acros
165131000 |
100 g | Glass bottle |
32.50 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13498398
|
Thermo Scientific Acros
165135000 |
500 g | Glass bottle |
107.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13448398
|
Thermo Scientific Acros
165130025 |
2.5 kg | Plastic bottle |
466.00 €
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
92-55-7 | |
243.17 | |
HSXKWKJCZNRMJO-UHFFFAOYSA-N | |
7097 | |
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C |
C9H9NO7 | |
MFCD00003244 | |
5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate | |
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate |
Specifications
92-55-7 | |
100.0 | |
Yellow | |
97.5% min. (GC) | |
MFCD00003244 | |
HSXKWKJCZNRMJO-UHFFFAOYSA-N | |
[acetyloxy-(5-nitrofuran-2-yl)methyl] acetate | |
7097 | |
98% | |
5-Nitro-2-furaldehyde diacetate, 98% |
97.5 | |
89.0°C to 93.0°C | |
Authentic | |
C9H9NO7 | |
5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetate | |
CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C | |
243.17 | |
243.17 | |
Fine Crystalline Powder |
Safety and Handling
missing translation for 'einecsNumber' : 202-166-1
missing translation for 'rtecsNumber' : LU1940000
missing translation for 'tsca' : TSCA
Product Identifier
- 5-Nitro-2-furaldehyde diacetate
RUO – Research Use Only