missing translation for 'onlineSavingsMsg'
Learn More
5-Nitro-2-furaldehyde diacetate, 98%, Thermo Scientific Chemicals Product Code.: 10791431

5-Nitro-2-furaldehyde diacetate, 98%, Thermo Scientific Chemicals

Product Code. 10791431
100 g, Glass bottle
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy

Product Code. 10791431

Brand: Thermo Scientific Acros 165131000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92-55-7
Molecular Formula C9H9NO7
Molecular Weight (g/mol) 243.17
MDL Number MFCD00003244
InChI Key HSXKWKJCZNRMJO-UHFFFAOYSA-N
Synonym 5-nitro-2-furaldehyde diacetate, 2-diacetoxymethyl-5-nitrofuran, 5-nitro-2-furanmethanediol diacetate, 5-nitrofurfural diacetate, 5-nitrofurfurylidene diacetate, 5-nitrofuran-2-yl methylene diacetate, nitrofuraldehyde diacetate, 5-nitro-2-furfural diacetate, 5-nitrofuraldehyde diacetate, 5-nitro-2-furfuraldehyde diacetateShow MoreShow Less
PubChem CID 7097
IUPAC Name [acetyloxy-(5-nitrofuran-2-yl)methyl] acetate
SMILES CC(=O)OC(C1=CC=C(O1)[N+](=O)[O-])OC(=O)C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 89.0°C to 93.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 100 g
Formula Weight 243.17
Percent Purity 98%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Nitro-2-furaldehyde diacetate, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0387141 Certificate TypeCertificate of Analysis Date12/12/2022 Catalog Number165130025,165131000,165135000
Lot NumberA0439192 Certificate TypeCertificate of Analysis Date17/02/2022 Catalog Number165130025,165131000,165135000
Product Identifier
  • 5-Nitro-2-furaldehyde diacetate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Nitro-2-furaldehyde diacetate, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.