missing translation for 'onlineSavingsMsg'
Learn More
5-Nitro-3-phenyl-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™ Product Code.: 10673413

5-Nitro-3-phenyl-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10673413
Amber Glass Bottle; 1g
Click to view available options
Amber Glass Bottle; 1g
This item is not returnable. View return policy

Product Code. 10673413

Brand: Thermo Scientific Maybridge RH00720DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 14182-37-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C15H10N2O4
MDL Number MFCD00054540
Quantity 1 g
Synonym 5-nitro-3-phenyl-1h-indole-2-carboxylic acid, maybridge1_006411, 1h-indole-2-carboxylicacid, 5-nitro-3-phenyl
InChI Key BMZNTPXQZKYUKD-UHFFFAOYSA-N
SMILES OC(=O)C1=C(C2=CC(=CC=C2N1)[N+]([O-])=O)C1=CC=CC=C1
IUPAC Name 5-nitro-3-phenyl-1H-indole-2-carboxylic acid
Molecular Weight (g/mol) 282.26
PubChem CID 700784
Formula Weight 282.25
Percent Purity 97%
Chemical Name or Material 5-Nitro-3-phenyl-1H-indole-2-carboxylic acid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
5-Nitro-3-phenyl-1H-indole-2-carboxylic acid, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.