missing translation for 'onlineSavingsMsg'
Learn More

5-Nitro-m-xylene, 99%, Thermo Scientific Chemicals

Product Code. 11312047
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

5-Nitro-m-xylene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Nitro-m-xylene was used in aerobic oxidation of aromatic anilines to aromatic azo compounds using gold nanoparticles supported on TiO2 as a catalyst.

Solubility
Insoluble in water.

Notes
Incompatible materials are Strong oxidizing agents, Strong bases, Strong reducing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-12-7
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.17
MDL Number MFCD00007269
InChI Key BYFNZOKBMZKTSC-UHFFFAOYSA-N
Synonym 5-nitro-m-xylene, 3,5-dimethylnitrobenzene, 3,5-dimethyl-1-nitrobenzene, benzene, 1,3-dimethyl-5-nitro, nitroxylol, m-xylene, 5-nitro, unii-5hvw2qtf0d, ccris 3121, 5hvw2qtf0d, 5-nitro-m-xylol
PubChem CID 7426
IUPAC Name 1,3-dimethyl-5-nitrobenzene
SMILES CC1=CC(=CC(C)=C1)[N+]([O-])=O

Specifications

Melting Point 69°C to 73°C
Boiling Point 273°C
Quantity 10 g
UN Number UN3447
Beilstein 1936130
Solubility Information Insoluble in water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 5-Nitro-m-xylene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'