missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 5-Nitroindoline, 97%

Product Code. 11497096
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 32692-19-6
Molecular Formula C8H8N2O2
Molecular Weight (g/mol) 164.164
MDL Number MFCD00005709
InChI Key WJQWYAJTPPYORB-UHFFFAOYSA-N
Synonym 5-nitroindoline, 1h-indole, 2,3-dihydro-5-nitro, indoline, 5-nitro, 2,3-dihydro-5-nitro-1h-indole, ccris 3306, 5-nitro-2,3-dihydro-indole, 5-nitroindolin, 5-nitro indoline, pubchem7442, acmc-1aera
PubChem CID 36219
IUPAC Name 5-nitro-2,3-dihydro-1H-indole
SMILES C1CNC2=C1C=C(C=C2)[N+](=O)[O-]

Specifications

Melting Point 92°C to 94°C
Quantity 5 g
Formula Weight 164.16
Percent Purity 97%
Chemical Name or Material 5-Nitroindoline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'