missing translation for 'onlineSavingsMsg'
Learn More

5-Nitrothiophene-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10009441
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 40357-96-8
Molecular Formula C5H3NO4S
Molecular Weight (g/mol) 173.142
MDL Number MFCD00052582
InChI Key VNJUNWUOAKEIKG-UHFFFAOYSA-N
Synonym 5-nitrothiophene-3-carboxylic acid, 2-nitrothiophene-4-carboxylic acid, 5-nitro-thiophene-3-carboxylic acid, 5-nitro-3-thiophenecarboxylic acid, 3-thiophenecarboxylic acid, 5-nitro, maybridge1_003249, acmc-1ascr, 2-nitro-4-carboxythiophene, ksc238c5f, 2-nitro-4-thiophenecarboxylic acid
PubChem CID 2777267
SMILES C1=C(SC=C1C(=O)O)[N+](=O)[O-]

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 173.15
Percent Purity 97%
Chemical Name or Material 5-Nitrothiophene-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'