missing translation for 'onlineSavingsMsg'
Learn More

5-Nonanone, 98%, Thermo Scientific Chemicals

Product Code. 10551601
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 502-56-7
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.24
MDL Number MFCD00009484
InChI Key WSGCRAOTEDLMFQ-UHFFFAOYSA-N
Synonym 5-nonanone, dibutyl ketone, butyl ketone, 5-oxononane, di-n-butyl ketone, unii-v8b2y1bz3s, hsdb 921, v8b2y1bz3s, n-c4h9 2co, dibutylketon
PubChem CID 10405
IUPAC Name nonan-5-one
SMILES CCCCC(=O)CCCC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -50.0°C
Color Colorless to Yellow
Density 0.8200g/mL
Boiling Point 186.0°C to 187.0°C
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)3CO(CH2)3CH3
Refractive Index 1.4180 to 1.4200
Quantity 100 mL
Beilstein 01, I, 365
Specific Gravity 0.82
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol;, very soluble in diethyl ether, chloroform.
Formula Weight 142.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 5-Nonanone, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.