missing translation for 'onlineSavingsMsg'
Learn More
Learn More
(5-Phenyl-1,3-oxazol-4-yl)methanol, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 175.19 |
Percent Purity | 95% |
Chemical Name or Material | (5-Phenyl-1, 3-oxazol-4-yl)methanol |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'