missing translation for 'onlineSavingsMsg'
Learn More

5-Phenyl-2-furaldehyde, 96%, Thermo Scientific Chemicals

176.00 € - 182.00 €

Chemical Identifiers

CAS 13803-39-9
Molecular Formula C11H8O2
Molecular Weight (g/mol) 172.19
InChI Key BMJHNNPEPBZULA-UHFFFAOYSA-N
Synonym 5-phenyl-2-furaldehyde, 2-formyl-5-phenylfuran, 5-phenyl-furan-2-carbaldehyde, 5-phenylfurfural, 2-furancarboxaldehyde,5-phenyl, 5-phenylfuraldehyde, acmc-209y7p, 5-phenyl-2-furancarboxaldehyde
PubChem CID 2769420
IUPAC Name 5-phenylfuran-2-carbaldehyde
SMILES C1=CC=C(C=C1)C2=CC=C(O2)C=O
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10159769
View Documents
Thermo Scientific Acros
437310010
1 g Glass bottle
176.00 €
1g
Please to purchase this item. Need a web account? Register with us today!
10844531
View Documents
Thermo Scientific Acros
437310050
5 g Glass bottle This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13803-39-9
172.19
5-phenyl-2-furaldehyde, 2-formyl-5-phenylfuran, 5-phenyl-furan-2-carbaldehyde, 5-phenylfurfural, 2-furancarboxaldehyde,5-phenyl, 5-phenylfuraldehyde, acmc-209y7p, 5-phenyl-2-furancarboxaldehyde
5-phenylfuran-2-carbaldehyde
C11H8O2
BMJHNNPEPBZULA-UHFFFAOYSA-N
2769420
C1=CC=C(C=C1)C2=CC=C(O2)C=O

Specifications

13803-39-9
100.0
Brown to Yellow
110°C
95% min. (GC)
5-phenyl-2-furaldehyde, 2-formyl-5-phenylfuran, 5-phenyl-furan-2-carbaldehyde, 5-phenylfurfural, 2-furancarboxaldehyde,5-phenyl, 5-phenylfuraldehyde, acmc-209y7p, 5-phenyl-2-furancarboxaldehyde
C1=CC=C(C=C1)C2=CC=C(O2)C=O
172.19
172.19
Liquid or Solid
95.0
29°C to 33°C
104°C to 108°C (6.0 mmHg)
Authentic
C11H8O2
BMJHNNPEPBZULA-UHFFFAOYSA-N
5-phenylfuran-2-carbaldehyde
2769420
96%
5-Phenyl-2-furaldehyde
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Warning

Product Identifier
  • 5-Phenyl-2-furaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.