missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phenyloxazole, Thermo Scientific™
86.20 € - 252.00 €
Specifications
Alpha Vector | PHENYLOXAZOLE |
---|---|
Chemical Name or Material | 5-Phenyloxazole |
CAS | 1006-68-4 |
Color | Brown to Yellow |
Infrared Spectrum | Conforms |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17740629
|
Thermo Scientific Acros
264220010 |
1 g |
86.20 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17750629
|
Thermo Scientific Acros
264220050 |
5 g |
252.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Specifications
PHENYLOXAZOLE | |
1006-68-4 | |
Conforms | |
C9 H7 N O | |
145.16 |
5-Phenyloxazole | |
Brown to Yellow | |
Glass bottle | |
00067074 | |
Crystals or Low Melting Solid |
Safety and Handling
missing translation for 'shelfLife' : 5 years
Recommended Storage : Normal conditions
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title