missing translation for 'onlineSavingsMsg'
Learn More

5-Phenylpyridine-2-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10402664
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 75754-04-0
Molecular Formula C12H9NO2
Molecular Weight (g/mol) 199.209
MDL Number MFCD04114189
InChI Key CNCJXSKAKGXNKJ-UHFFFAOYSA-N
Synonym 5-phenylpicolinic acid, 2-pyridinecarboxylic acid, 5-phenyl, 5-phenylpicolinicacid, pubchem22107, 5-phenyl-picolinic acid, acmc-1bb5a, 2-carboxy-5-phenylpyridine, 3-phenyl-6-carboxypyridine, 5-phenyl-2-pyridinecarboxylic acid, 5-phenyl-pyridine-2-carboxylic acid
PubChem CID 2762870
IUPAC Name 5-phenylpyridine-2-carboxylic acid
SMILES C1=CC=C(C=C1)C2=CN=C(C=C2)C(=O)O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 199.21
Percent Purity 95%
Chemical Name or Material 5-Phenylpyridine-2-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.