missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Phenylthiophene-2-carboxylic acid, 95+%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Formula Weight | 204.25 |
Percent Purity | 95% |
Chemical Name or Material | 5-Phenylthiophene-2-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'