missing translation for 'onlineSavingsMsg'
Learn More
Learn More
5-Pyrimidinylboronic acid, 97%, Thermo Scientific™
17.70 € - 291.00 €
Chemical Identifiers
CAS | 109299-78-7 |
---|---|
Molecular Formula | C4H5BN2O2 |
Molecular Weight (g/mol) | 123.91 |
MDL Number | MFCD03002366 |
InChI Key | HZFPPBMKGYINDF-UHFFFAOYSA-N |
Synonym | 5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid |
PubChem CID | 2795193 |
SMILES | OB(O)C1=CN=CN=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10147712
|
Thermo Scientific Maybridge
CC07412CB |
250 mg | Amber Glass Bottle |
17.70 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10284062
|
Thermo Scientific Maybridge
CC07412DA |
1 g | Amber Glass Bottle |
56.40 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10624765
|
Thermo Scientific Maybridge
CC07412EA |
10 g | Amber Glass Bottle |
291.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
109299-78-7 | |
123.91 | |
HZFPPBMKGYINDF-UHFFFAOYSA-N | |
2795193 |
C4H5BN2O2 | |
MFCD03002366 | |
5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid | |
OB(O)C1=CN=CN=C1 |
Specifications
109299-78-7 | |
C4H5BN2O2 | |
5-pyrimidinylboronic acid, pyrimidine-5-boronic acid, 5-pyrimidineboronic acid, 5-pyrimidylboronic acid, pyrimidin-5-yl boronic acid, 5-pyrimidine boronic acid, pyrimidinyl-5-boronic acid, pyrimidine-5-ylboronic acid, pyrimidin-5-yl-5-boronic acid, pyrimidin-5-boronic acid | |
OB(O)C1=CN=CN=C1 | |
123.91 | |
123.91 | |
5-Pyrimidinylboronic acid |
97% | |
MFCD03002366 | |
HZFPPBMKGYINDF-UHFFFAOYSA-N | |
pyrimidin-5-ylboronic acid | |
2795193 | |
97% |
Safety and Handling
Warning