missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfoisophthalic acid monosodium salt, 95%, Thermo Scientific Chemicals

Product Code. 11334649
Click to view available options
Quantity:
100 g
250 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product's dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It can be used as the third monomer in the Polyester procession of esterification, which can improve the ability of product′s dying. And it can be used as the catalyst in medicine, agricultural chemical, polyester. Used as a monomer for waterborne polyesters and nylon fibers, in adhesion aid for magnetic tapes, and amine free polyurethane dispersions.

Solubility
Soluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6362-79-4
Molecular Formula C8H5NaO7S
Molecular Weight (g/mol) 268.171
MDL Number MFCD00007495
InChI Key YXTFRJVQOWZDPP-UHFFFAOYSA-M
Synonym 5-sulfafe-3-phthalicacid
PubChem CID 57488606
IUPAC Name sodium;3-carboxy-5-sulfobenzoate
SMILES C1=C(C=C(C=C1C(=O)O)S(=O)(=O)O)C(=O)[O-].[Na+]

Specifications

Melting Point >300°C
Quantity 100 g
Beilstein 4170770
Solubility Information Soluble in water.
Formula Weight 268.18
Percent Purity 95%
Chemical Name or Material 5-Sulfoisophthalic acid monosodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'