missing translation for 'onlineSavingsMsg'
Learn More

5-Sulfosalicylic Acid Dihydrate, 98%, Thermo Scientific Chemicals

Brand:  Thermo Scientific Acros 132860010

101.05 EUR valid until 2024-12-31
Use promo code "21665" to get your promotional price.



 View more versions of this product

Product Code. 10644612

  • 111.56 € / 1kg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5-Sulfosalicylic acid dihydrate
97.5
106.0°C to 110.0°C
Authentic
Plastic bottle
MFCD00007508,MFCD00149540
11, 411
15, 9094
Solubility in water: 127g/L (20°C). Other solubilities: soluble in alcohol and ether
C.C.OC(=O)C1=CC(=CC=C1O)S(O)(=O)=O
250.27
254.22
Crystalline Powder or Crystals
5965-83-3, 97-05-2
100.0
White
98%
C9H14O6S
1 kg
01,1118
2-hydroxy-5-sulfobenzoic acid dihydrate, 5-sulfosalicylic acid dihydrate, sulfosalicylic acid dihydrate, benzoic acid, 2-hydroxy-5-sulfo-, dihydrate, unii-09ngq462s6, 2-hydroxy-5-sulfobenzoic acid, hydrate, hydrate, 5-sulfosalicylsyre, 5-sulfosalicylsaeure, salicylic acid, 5-sulfo-, dihydrate, acmc-1aymd
NFYHZVWMQHQKRU-UHFFFAOYSA-N
2-hydroxy-5-sulfobenzoic acid;dihydrate
2723734
98%
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 5-Sulfosalicylic acid dihydrate

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

Supplemental information
  • MIXTURE LIST-Contains : 5-Sulphosalicylic acid dihydrate

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

RUO – Research Use Only