missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Probes™ 5-TAMRA, SE (5-Carboxytetramethylrhodamine, Succinimidyl Ester), single isomer
5-TAMRA, SE (5-Carboxytetramethylrhodamine, Succinimidyl Ester), single isomer
Brand: Molecular Probes™ C2211
500.08 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
The amine-reactive 5-TAMRA, SE and its conjugates yield bright, pH-insensitive orange-red fluorescence (approximate excitation/emission maxima ∼546/579) with good photostability.Specifications
5 mg | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature | |
TAMRA™ Isomers, TMR (Tetramethylrhodamine) | |
579 |
5-TAMRA SE | |
Amine | |
Active Ester, Succinimidyl Ester | |
Classic Dyes | |
546 |
For Research Use Only. Not for use in diagnostic procedures.