missing translation for 'onlineSavingsMsg'
Learn More

5-(tert-Butyl)-3-(chloromethyl)-1,2,4-oxadiazole, 95+%, Thermo Scientific™

Product Code. 10099521
Click to view available options
Quantity:
250 mg
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10099521

Brand: Thermo Scientific Maybridge SEW00330DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175205-41-1
Molecular Formula C7H11ClN2O
Molecular Weight (g/mol) 174.628
InChI Key AIVSLDAXUIRICJ-UHFFFAOYSA-N
PubChem CID 2796738
IUPAC Name 5-tert-butyl-3-(chloromethyl)-1,2,4-oxadiazole
SMILES CC(C)(C)C1=NC(=NO1)CCl

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 5-tert-butyl-3-chloromethyl-1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl-1,2,4-oxadiazole, 1,2,4-oxadiazole,3-chloromethyl-5-1,1-dimethylethyl, 3-chloromethyl-5-t-butyl-1,2,4-oxadiazole, 3-chloromethyl-5-tert-butyl-1,2,4-oxadiazole, 1,2,4-oxadiazole, 3-chloromethyl-5-1,1-dimethylethyl
Formula Weight 174.63
Percent Purity 95%
Chemical Name or Material 5-(tert-Butyl)-3-(chloromethyl)-1, 2, 4-oxadiazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.