missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcon 50 mL ViewPoint™ Centrifuge Thermochromic Tubes
Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
19.90 € - 60.10 €
Specifications
Certifications/Compliance | ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity |
---|---|
Closure Color | Purple |
Color | Clear to Blue color change at 0°C to 4°C |
Capacity (Metric) | 50 mL |
Disposable | Single-use |
Product Code | Brand | No. per Pack | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. per Pack | Sterility | Price | Quantity & Availability | ||||
16639842
|
Labcon
3220-320-006-9 |
10/IntegraPack™ Bag | Non-sterile |
59.45 €
Pack of 50 |
Estimated Shipment: 19-11-2024
Log in to see stock. |
||||
16649842
|
Labcon
3220-325-006-9 |
10/IntegraPack™ Bag | Sterile | ||||||
16659842
|
Labcon
3220-335-008-9 |
25/Rack | Sterile |
19.90 €
Pack of 25 |
In Stock
Log in to see stock. |
||||
Description
- Real-time temperature signaling
- Reversibly transition from clear to light blue at 4°C with full dark blue transition at 0°C
- With attached flat screw caps
- Black graduations and white writing surface
- ATP, RNase/DNase and Endotoxin Free (non-pyrogenic)
- Sperm Motility Assay (SMA) Mouse Embryo Assay (MEA) validated safe for cell culture
- Polypropylene USP Class VI resin
- Centrifuging to 12,500 x G
- Freezable to -80°C
- Made in USA
Specifications
ISO 9001:2015; FDA CFR21 GMP; CE IVD Conformity | |
Clear to Blue color change at 0°C to 4°C | |
Single-use | |
Room Temperature | |
115 mm | |
Conical | |
29 mm | |
Conical Centrifuge Tube |
Purple | |
50 mL | |
ViewPoint™ | |
Autoclavable | |
29 mm | |
Yes | |
5 to 45 in increments of 1 | |
12,500 x g |