missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ 5000 Series 0.03μm to 0.30μm Polymer Particle Suspensions

Product Code. 11819802 Shop All Thermo Scientific Products
Click to view available options
Diameter (Metric):
0.03 μm
0.06 μm
0.08 μm
0.09 μm
0.10 μm
0.11 μm
0.12 μm
0.14 μm
0.16 μm
0.17 μm
0.20 μm
0.22 μm
0.24 μm
0.26 μm
0.3 μm
Quantity:
15 mL
100 mL
1000 mL
Unit Size:
1000mL
100mL
15mL
This item is not returnable. View return policy

Product Code. 11819802

Brand: Thermo Scientific™ 5009B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Optimize research in light scattering, microporous filter checking, and aerosol particle generation with these particle suspensions.

Optimize research in light scattering, microporous filter checking, and aerosol particle generation with Thermo Scientific™ 5000 Series 0.03μm to 0.30μm Polymer Particle Suspensions. These particles are designed to meet the need for particulate materials with a variety of particle sizes and properties.

The polystyrene microspheres have a density of 1.05g/cm3 and a refractive index of 1.59 at 589nm. Particle diameters are measured by optical microscopy, photon correlation spectroscopy or light scattering. They are packaged as aqueous suspensions at 10% solids by weight. The 5000 Series particles range up to 3.1 microns in size. If you require larger particles, please refer to our 7000 Series Copolymer Microsphere Suspensions.

Ordering Information:

15mL, 100mL, and 1000mL bottle sizes available. 15mL (“A” Part No. bottles), 100mL (B bottles) and 1000mL (C bottles) are packaged to order; example (Part No. 5003B and 5003C). Please allow 10 working days when ordering these fill volumes.

Includes:

Material Safety Data Sheet (MSDS) and general product handling insert sheet

Recommended for:

High concentration polymer particles < 3μm used in filter evaluation and testing, fluid mechanics research, dispersion studies and many other research and development projects. Not intended for use in instrument calibration or diagnostic reagents because they lack the exacting specifications needed for those applications.

TRUSTED_SUSTAINABILITY

Specifications

Additive Contains trace amount of surfactant to inhibit agglomeration and promote stability
Concentration 10%
Content And Storage Unless otherwise stated, refrigerate (2° to 8°C) product when not in use but do not freeze. Store upright and keep bottle tightly sealed. Mix product with gentle inversion by hand or vortex mixer.
Description Particles useful for such applications as filter evaluation, checking and testing, light scattering research, fluid mechanics research, aerosol particle generation, dispersion studies, and many other research and development projects. They are not intended for use in instrument calibration.
Color White or Colorless
For Use With (Application) Aerosol Particle Generation, Dispersion, Filter Evaluation, Light Scattering, Particle Generation
Material Polystyrene
Product Type Polymer Particle Suspensions
Includes General product handling insert sheet
Packaging 100 mL
Particle Density 1.05g/cm3
Quantity 100 mL
Refractive Index 1.59 at 589nm (25°C)
Stability Product stable for a minimum of 24 months
Diameter (Metric) 0.09 μm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.