missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Spectrum™ Labs Spectra/Por™ 7 8000 D MWCO Standard RC Pre-Treated Dialysis Kits
Click to view available options
Diameter (Metric):
7.5 mm
15 mm
20.4 mm
25.5 mm
32 mm
Length (Metric):
0.45 mL/cm vol./length
1.8 mL/cm vol./length
3.3 mL/cm vol./length
5.1 mL/cm vol./length
7.9 mL/cm vol./length
Unit Size:
1m
Description
Includes
- 1meter of pre-treated tubing, a pair of closures and 5 opening picks
- Hydrophilic, symmetric porositiy
- Equilibrated in water
- Packaged wet in preservative solution of 0.05% sodium azide
- For small experiments or qualifying a membrane for a particular application
Specifications
Specifications
| MWCO | 8 kDa |
| Diameter (Metric) | 25.5 mm |
| Length (Metric) | 5.1 mL/cm vol./length |
| Quantity | 1 m Tubing/Kit |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction