missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ iBind™ Flex Western Device
Allows better antigen-antibody interaction and solutions flow between the iBind™ Card and membrane for maximum interaction between protein and antibody
2465.00 € - 4185.00 €
Specifications
Capacity | 1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots |
---|---|
Detection Method | Fluorescence, Chemiluminescence, Chromogenic |
For Use With (Application) | Western Blot |
Membrane Compatibility | Nitrocellulose, PVDF |
Product Line | iBind Flex |
Product Code | Brand | dimProductType | KitContents | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | dimProductType | KitContents | Price | Quantity & Availability | ||||
15357268
|
Invitrogen™
SLF2000 |
2465.00 €
Each |
In Stock
Log in to see stock. |
||||||
17134609
|
Invitrogen™
SLF20002PK |
4185.00 €
Pack of 2 |
In Stock
Log in to see stock. |
||||||
Description
The iBind™ Flex Western System offers automated western blot processing and can accommodate various blot formats including midi, mini, and vertically cut strips. Based on Sequential Lateral Flow (SLF) technology, all blocking, antibody incubation and washing steps during immunodetection are hands-free. Compared to manual methods, the iBind Flex Western System requires less primary antibody, enables reproducible results and is compatible with downstream detection protocols.The Invitrogen™ iBind™ Flex Western System offers:
- Flexible options—optimize antibody use, change formats and reduce hands-on time
- Cost savings—use up to 80% less primary antibody
- Reproducibility—automated processing provides improved blot-to-blot consistency
Specifications
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots | |
Western Blot | |
iBind Flex | |
1 Midi Blot, 6 Vertically-Cut Strip Blots, 2 Mini Blots | |
Room Temperature |
Fluorescence, Chemiluminescence, Chromogenic | |
Nitrocellulose, PVDF | |
∼2.5 hr. | |
Western Device |
For Research Use Only. Not for use in diagnostic procedures.