Learn More
Spectrum™ Labs Spectra/Por™ 1 6-8 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
48.67 € - 279.00 €
Specifications
MWCO | 6 to 8 kDa |
---|
Includes:
- 1m or 5m of Spectra/Por 1 dialysis tubing
- 1 Standard Spectra/Por Closure (for buoyance)
- 1 weighted Spectra/Por Closure (for orientation)
- 2 adhesive tubing openers
Product Code | Brand | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Quantity | Price | Quantity & Availability | |||
15370752
|
Spectrum™
132675T |
76 mm | 46 mL/cm vol./length | 5 m Tubing/Kit |
259.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15350752
|
Spectrum™
132670T |
64 mm | 32 mL/cm vol./length | 5 m Tubing/Kit |
245.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15380752
|
Spectrum™
132 675T1 |
76 mm | 46 mL/cm vol./length | 1 m Tubing/Kit | N/A | ||||
15330752
|
Spectrum™
132 665T1 |
32 mm | 7.9 mL/cm vol./length | 1 m Tubing/Kit | N/A | ||||
15360752
|
Spectrum™
132 670T1 |
64 mm | 32 mL/cm vol./length | 1 m Tubing/Kit | N/A | ||||
15320752
|
Spectrum™
132 660T1 |
25.5 mm | 5.1 mL/cm vol./length | 1 m Tubing/Kit | N/A | ||||
15300752
|
Spectrum™
132 655T1 |
20.4 mm | 3.3 mL/cm vol./length | 1 m Tubing/Kit | N/A | ||||
15310752
|
Spectrum™
132660T |
25.5 mm | 5.1 mL/cm vol./length | 5 m Tubing/Kit |
279.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15380742
|
Spectrum™
132 650T1 |
14.6 mm | 1.7 mL/cm | 1 m Tubing/Kit | N/A | ||||
15370742
|
Spectrum™
132650T |
14.6 mm | 1.7 mL/cm | 5 m Tubing/Kit |
239.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15350742
|
Spectrum™
132645T |
6.4 mm | 0.32 mL/cm vol./length | 5 m Tubing/Kit |
233.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15360742
|
Spectrum™
132 645T1 |
6.4 mm | 0.32 mL/cm vol./length | 1 m Tubing/Kit | N/A | ||||
11521040
|
Spectrum™
132665T |
32 mm | 5 m | Each |
257.00 €
5m |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- For basic dialysis applications, or evaluating dialysis tubing for dialysis application
- Packaged dry with glycerin (soak in water)
Specifications
6 to 8 kDa |