Learn More
Spectrum™ Labs Spectra/Por™ 2 12-14 kD MWCO Standard RC Dry Dialysis Kits
With Standard Grade Regenerated Cellulose membrane tubing for applications with significant size difference between MW species being separated
Brand:  Spectrum™ 132682T
 View more versions of this product
Product Code. 15340762
  • 282.00 € / 5m
Explore more special offers
Includes:
  • 1m or 5m of Spectra/Por 2 dialysis tubing
  • 1 Standard Spectra/Por Closure (for buoyance)
  • 1 weighted Spectra/Por Closure (for orientation)
  • 2 adhesive tubing openers
Description
Description
  • For higher permeability applications, or evaluating dialysis tubing for dialysis application
  • Packaged dry with glycerin (soak in water)
Specifications
Specifications
12 to 14 kDa
67 mm
34 mL/cm vol./length
5 m Tubing/Kit