Learn More
Cellpath Stainless-steel Reusable Base Molds
Product Code. p-5046819
Click to view available options
Dimensions (L x W x H):
7 x 7 x 5 mm
10 x 24 x 5 mm
14 x 24 x 5 mm
24 x 24 x 5 mm
30 x 24 x 9 mm
37 x 24 x 9 mm
Unit Size:
Pack of 5
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 30226270
Brand: Cellpath GBB071405A
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Ideal for prostrate and breast cores and can be used with both grid and slatted processing cassettes for both horizontal and vertical sectioning. Cellpath Stainless-steel Reusable Base Moulds are manufactured from high quality stainless steel that put an end to extensive trimming of wax.
  • Made of reusable Stainless steel
  • Stackable to reduce storage area
  • Internal rounded corners enable easy removal of the wax block
  • Features extended edges prevent tipping and provide grip area for ease of handling
  • Available in wide range of sizes
  • GBB-0714-05A suitable for use with needle and punch biopsies
Specifications
Material Stainless Steel
Dimensions (L x W x H) 7 x 7 x 5 mm
Quantity 5/Pk.