Learn More
Eppendorf™ Rotor for Centrifuge 5810 / 5810 R
Eppendorf Rotor for Centrifuge 5810/5810 R - precision redefined. High-capacity centrifugation with varied rotor options, a range of capacities and speeds, and versatile buckets to ensure efficient sample processing. Ideal for diverse applications, guaranteeing accuracy and reproducibility.
1580.00 € - 3385.00 €
Specifications
Type Rotor
For Use With (Equipment) Centrifuge 5810 / 5810 R
View More Specs
Products 4
Product Code Brand Description Capacity Max. RCF Max. Speed Price Quantity & Availability  
Product Code Brand Description Capacity Max. RCF Max. Speed Price Quantity & Availability  
12806233
View Documents
Eppendorf™
5820740000
S-4-104 - Includes 750 mL round buckets (x4) 4 x 750 mL 3,214 x g 3,900 rpm
3385.00 €
Each
15214536
missing translation for 'viewDocuments'
Eppendorf™
5820755008
S-4-104 - without buckets 4 x 750 mL 3,214 x g 3,900 rpm
1580.00 €
Each
12876233
missing translation for 'viewDocuments'
Eppendorf™
5820754001
S-4-104 - Includes plate buckets (x4) 4 x 750 mL 3,214 x g 3,900 rpm
3385.00 €
Each
10626250
missing translation for 'viewDocuments'
Eppendorf™
5820710004
A-2-DWP-AT - Includes buckets (x2), aerosol-tight caps (x2), plate carriers (x2) 2 x 4 MTP, 2 x 1 DWP 3,486 x g 4,500 rpm
2680.00 €
Each
Beschreibung
Beschreibung
  • Precision-engineered for optimal sample separation.
  • All fixed-angle rotors, rotor lids, buckets, caps and adapters can be autoclaved (121°C, 20 min.).
  • Accessories (adaptors and buckets) further customize your centrifuge setup.
Spezifikation
Spezifikation
Rotor
Centrifuge 5810 / 5810 R
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate